Search
Search

| Assay Name: | hsa-miR-554 |
|---|---|
| miRBase Accession Number: | MI0003559 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCUAGUCCUGACUCAGCCAGU |
| Species: | Human, Pan troglodytes, Rhesus monkey |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000551, mir-554 |
Mature miRNA Sequence: | GCUAGUCCUGACUCAGCCAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-554 | MIMAT0003217 |
| Pan troglodytes | ptr-miR-554 | MIMAT0008235 |
| Rhesus monkey | mml-miR-554 | MIMAT0006423 |