Search
Search

| Assay Name: | rlcv-miR-rL1-14-5p |
|---|---|
| miRBase Accession Number: | MI0003750 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCGGACGGUCUGGUGCGCUUGAU |
| Species: | Rhesus lymphocryptovirus |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000993, mir-rL1-14 |
Mature miRNA Sequence: | UCGGACGGUCUGGUGCGCUUGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rhesus lymphocryptovirus | rlcv-miR-rL1-14-5p | MIMAT0003444 |