Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12495
          Assay Name: hsa-miR-889-3p
          miRBase Accession Number: MI0005540
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101047901 - 101047979 [+] on Build GRCh38
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Species: Human, Callithrix jacchus, Dasypus novemcinctus, Guinea pig, Horse, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12495

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000514, mir-889

          Mature miRNA Sequence:

          UUAAUAUCGGACAACCAUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-889-3p MIMAT0004921
          Callithrix jacchus cja-miR-889 MIMAT0049325
          Dasypus novemcinctus dno-miR-889-3p MIMAT0048023
          Guinea pig cpo-miR-889-3p MIMAT0047398
          Horse eca-miR-889 MIMAT0013175
          Pan paniscus ppa-miR-889 MIMAT0049039
          View More Rows
          Pan troglodytes ptr-miR-889 MIMAT0008331
          Pongo pygmaeus ppy-miR-889 MIMAT0016124
          Pteropus alecto pal-miR-889-3p MIMAT0040227
          Rabbit ocu-miR-889-3p MIMAT0048582
          Rhesus monkey mml-miR-889-3p MIMAT0006528

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-889
          Stem-loop Accession # MI0005540
          Stem-loop Sequence
          GUGCUUAAAGAAUGGCUGUCCGUAGUAUGGUCUCUAUAUUUAUGAUGAUUAAUAUCGGACAACCAUUGUUUUAGUAUCC
          Chromosome Location Chr. 14 - 101047901 - 101047979 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004921
          miRBase ID: hsa-miR-889-3p
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: Chr. 14 - 101047901 - 101047979 [+] on Build GRCh38

          Callithrix jacchus

          Stem-loop ID cja-mir-889
          Stem-loop Accession # MI0040061
          Stem-loop Sequence
          AAUGGCUGUCCGUGGUAUGGUCUCUCUAUUUAUGAUGAUUAAUAUCGGACAACCAUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049325
          miRBase ID: cja-miR-889
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-889
          Stem-loop Accession # MI0039178
          Stem-loop Sequence
          AAUGGCUGUCCGUAGUAUGGUCUCUAUAUUUAUGAUGAUUAAUAUCGGACAACCAUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048023
          miRBase ID: dno-miR-889-3p
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-889
          Stem-loop Accession # MI0038848
          Stem-loop Sequence
          AAUGGCUGUCCGUAGUGUGGUCUCUAUAUUUACGAUGAUUAAUAUCGGACAACCAUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047398
          miRBase ID: cpo-miR-889-3p
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-889
          Stem-loop Accession # MI0012918
          Stem-loop Sequence
          GUGCUUAAAGAAUGGCUGUCUGUAGUAUGGUCUCCAUAUUCAUGGUGAUUAAUAUCGGACAACCAUUGUUUUAGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013175
          miRBase ID: eca-miR-889
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-889
          Stem-loop Accession # MI0039747
          Stem-loop Sequence
          AAUGGCUGUCCGUAGUAUGGUCUCUAUAUUUAUGAUGAUUAAUAUCGGACAACCAUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049039
          miRBase ID: ppa-miR-889
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-889
          Stem-loop Accession # MI0008870
          Stem-loop Sequence
          UGCUUAAAGAAUGGCUGUCCGUAGUAUGGUCUCUAUAUUUAUGAUGAUUAAUAUCGGACAACCAUUGUUUUAGUAUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008331
          miRBase ID: ptr-miR-889
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-889
          Stem-loop Accession # MI0015159
          Stem-loop Sequence
          GUGCUUAAAGAAUGGCUGUCCGUAGUAUGGUCUCUAUAUUUAUGAUGAUUAAUAUCGGACAACCAUUGUUUUAGUAUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016124
          miRBase ID: ppy-miR-889
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-889
          Stem-loop Accession # MI0032561
          Stem-loop Sequence
          AAUGGCUGUCCGUAGUAUGGUCUCUAUAUUUACAGUGAUUAAUAUCGGACAACCAUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040227
          miRBase ID: pal-miR-889-3p
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-889
          Stem-loop Accession # MI0039473
          Stem-loop Sequence
          AAUGGCUGUCCGUAGUAUGGUCUCUAUAUUUAUGGUGAUUAAUAUCGGACAACCAUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048582
          miRBase ID: ocu-miR-889-3p
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-889
          Stem-loop Accession # MI0007923
          Stem-loop Sequence
          GUGCUUAAAGAAUGGCUGUCCGUAGUAUGGUCUCUAUAUUUAUGAUGAUUAAUAUCGGACAACCAUUGUUUUAGUAUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006528
          miRBase ID: mml-miR-889-3p
          Mature miRNA Sequence: UUAAUAUCGGACAACCAUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305121_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12495
          Ambion® Pre-miR™ miRNA Precursor : PM12495
          TaqMan™ MicroRNA Assay : 002202
          mirVana® miRNA inhibitor : MH12495
          TaqMan™ Advanced miRNA Assay : 478208_mir


          miRBase Alias:

          Human: hsa-miR-889 (v19)
          Macaca mulatta: mml-miR-889 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline