Search
Search

| Assay Name: | hsa-miR-339-3p |
|---|---|
| miRBase Accession Number: | MI0000815 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGCGCCUCGACGACAGAGCCG |
| Species: | Human, Callithrix jacchus, Daubentonia madagascariensis, Gorilla gorilla, Rhesus monkey |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000193, mir-339 |
Mature miRNA Sequence: | UGAGCGCCUCGACGACAGAGCCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-339-3p | MIMAT0004702 |
| Callithrix jacchus | cja-miR-339 | MIMAT0039482 |
| Daubentonia madagascariensis | dma-miR-339 | MIMAT0049299 |
| Gorilla gorilla | ggo-miR-339 | MIMAT0024161 |
| Rhesus monkey | mml-miR-339-3p | MIMAT0006280 |