Search
Search

| Assay Name: | mmu-miR-342-5p |
|---|---|
| miRBase Accession Number: | MI0000627 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGGGUGCUAUCUGUGAUUGAG |
| Species: | Mouse, Rat, Horse |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000190, mir-342 |
Mature miRNA Sequence: | AGGGGUGCUAUCUGUGAUUGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-342-5p | MIMAT0004653 |
| Rat | rno-miR-342-5p | MIMAT0004652 |
| Horse | eca-miR-342-5p | MIMAT0013136 |