Search
Search

| Assay Name: | hsa-miR-636 |
|---|---|
| miRBase Accession Number: | MI0003651 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUGCUUGCUCGUCCCGCCCGCA |
| Species: | Human |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000524, mir-636 |
Mature miRNA Sequence: | UGUGCUUGCUCGUCCCGCCCGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-636 | MIMAT0003306 |