Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13067
          Assay Name: hsa-miR-935
          miRBase Accession Number: MI0005757
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 53982307 - 53982397 [+] on Build GRCh38
          Mature miRNA Sequence: CCAGUUACCGCUUCCGCUACCGC
          Species: Human, Rat, Bovine, Pan troglodytes, Pig
          Product Type: mirVana® miRNA mimic
          Assay ID MC13067

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000606, mir-935

          Mature miRNA Sequence:

          CCAGUUACCGCUUCCGCUACCGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-935 MIMAT0004978
          Rat rno-miR-935 MIMAT0012845
          Bovine bta-miR-935 MIMAT0009385
          Pan troglodytes ptr-miR-935 MIMAT0008342
          Pig ssc-miR-935 MIMAT0013947

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-935
          Stem-loop Accession # MI0005757
          Stem-loop Sequence
          GGCGGGGGCGCGGGCGGCAGUGGCGGGAGCGGCCCCUCGGCCAUCCUCCGUCUGCCCAGUUACCGCUUCCGCUACCGCCGCCGCUCCCGCU
          Chromosome Location Chr. 19 - 53982307 - 53982397 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004978
          miRBase ID: hsa-miR-935
          Mature miRNA Sequence: CCAGUUACCGCUUCCGCUACCGC
          Chromosome Location: Chr. 19 - 53982307 - 53982397 [+] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-935
          Stem-loop Accession # MI0012607
          Stem-loop Sequence
          GGCGGCGGCGCGGGCGGCAGUGGCGGGAGCGGCCCCUCGGCCAUCCUCCGUCUGCCCAGUUACCGCUUCCGCUACCGCCGCCGCUCCCGCU
          Chromosome Location Chr. 1 - 64330549 - 64330639 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0012845
          miRBase ID: rno-miR-935
          Mature miRNA Sequence: CCAGUUACCGCUUCCGCUACCGC
          Chromosome Location: Chr. 1 - 64330549 - 64330639 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-935
          Stem-loop Accession # MI0009908
          Stem-loop Sequence
          GGCGGGGGCGCGGGCGGCAGUGGCGGGAGCGGCCCCUCGGCCAUCCUCCGUCUGCCCAGUUACCGCUUCCGCUACCGCCGCCGCUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009385
          miRBase ID: bta-miR-935
          Mature miRNA Sequence: CCAGUUACCGCUUCCGCUACCGC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-935
          Stem-loop Accession # MI0008882
          Stem-loop Sequence
          GCGGGGGCGCGGGCGGCAGUGGCGGGAGCGGCCCCUCGGCCAUCCUCCGUCUGCCCAGUUACCGCUUCCGCUACCGCCGCCGCUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008342
          miRBase ID: ptr-miR-935
          Mature miRNA Sequence: CCAGUUACCGCUUCCGCUACCGC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-935
          Stem-loop Accession # MI0013156
          Stem-loop Sequence
          GGCGCAGGCGGCAGUGGCGGGAGCGGCCCCUCGGCCAUCCUCCGUCUGCCCAGUUACCGCUUCCGCUACCGCCGCCGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013947
          miRBase ID: ssc-miR-935
          Mature miRNA Sequence: CCAGUUACCGCUUCCGCUACCGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn04229323_pri
          TaqMan™ Pri-miRNA Assay : Hs03295745_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13067
          Ambion® Pre-miR™ miRNA Precursor : PM13067
          TaqMan™ MicroRNA Assay : 002178
          mirVana® miRNA inhibitor : MH13067
          TaqMan™ Advanced miRNA Assay : rno479210_mir
          TaqMan™ Advanced miRNA Assay : 479210_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline