Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13128
          Assay Name: hsa-miR-548f-3p
          miRBase Accession Number: MI0006374
          miRBase Version: v22.1
          Chromosome Location: Chr.10: 54607874 - 54607957 [-] on Build GRCh38
          Mature miRNA Sequence: AAAAACUGUAAUUACUUUU
          Species: Human, Pan troglodytes
          Product Type: mirVana® miRNA mimic
          Assay ID MC13128

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000317, mir-548

          Mature miRNA Sequence:

          AAAAACUGUAAUUACUUUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-548f-3p MIMAT0005895
          Pan troglodytes ptr-miR-548f MIMAT0008221

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-548f-1
          Stem-loop Accession # MI0006374
          Stem-loop Sequence
          AUUAGGUUGGUGCAAAAGUAAUCACAGUUUUUGACAUUACUUUCAAAGACAAAAACUGUAAUUACUUUUGGACCAACCUAAUAG
          Chromosome Location Chr. 10 - 54607874 - 54607957 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005895
          miRBase ID: hsa-miR-548f-3p
          Mature miRNA Sequence: AAAAACUGUAAUUACUUUU
          Chromosome Location: Chr. X - 32641474 - 32641559 [-] on Build GRCh38

          Stem-loop ID hsa-mir-548f-2
          Stem-loop Accession # MI0006375
          Stem-loop Sequence
          UAAUAACUAUUAGGUUGGUGCGAACAUAAUUGCAGUUUUUAUCAUUACUUUUAAUGGCAAAAACUGUAAUUACUUUUGCACCAACCUAAUAUUUUAGU
          Chromosome Location Chr. 2 - 212426263 - 212426360 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005895
          miRBase ID: hsa-miR-548f-3p
          Mature miRNA Sequence: AAAAACUGUAAUUACUUUU
          Chromosome Location: Chr. X - 32641474 - 32641559 [-] on Build GRCh38

          Stem-loop ID hsa-mir-548f-3
          Stem-loop Accession # MI0006376
          Stem-loop Sequence
          AUUAGGUUGGUGCAAACCUAAUUGCAAUUUUUGCAGUUUUUUUAAGUAAUUGCAAAAACUGUAAUUACUUUUGCACCAACCUAAUAC
          Chromosome Location Chr. 5 - 110513829 - 110513915 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005895
          miRBase ID: hsa-miR-548f-3p
          Mature miRNA Sequence: AAAAACUGUAAUUACUUUU
          Chromosome Location: Chr. X - 32641474 - 32641559 [-] on Build GRCh38

          Stem-loop ID hsa-mir-548f-4
          Stem-loop Accession # MI0006377
          Stem-loop Sequence
          GAGUUCUAACGUAUUAGGUUGGUGCAAAAGUAAUAGUGGUUUUUGCCAUUAAAAGUAAUGACAAAAACUGUAAUUACUUUUGGAACAAUAUUAAUAGAAUUUCAG
          Chromosome Location Chr. 7 - 147378017 - 147378121 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005895
          miRBase ID: hsa-miR-548f-3p
          Mature miRNA Sequence: AAAAACUGUAAUUACUUUU
          Chromosome Location: Chr. X - 32641474 - 32641559 [-] on Build GRCh38

          Stem-loop ID hsa-mir-548f-5
          Stem-loop Accession # MI0006378
          Stem-loop Sequence
          UAUUAGGUUGCUGCAAAAGUAAUCAUGUUUUUUUCCAUUGUAAGUAAUGGGAAAAACUGUAAUUACUUUUGUACCAACCUAAUAGC
          Chromosome Location Chr. X - 32641474 - 32641559 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005895
          miRBase ID: hsa-miR-548f-3p
          Mature miRNA Sequence: AAAAACUGUAAUUACUUUU
          Chromosome Location: Chr. X - 32641474 - 32641559 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-548f-1
          Stem-loop Accession # MI0008750
          Stem-loop Sequence
          UUAGGUUGGUGCAAAAGAAAUCAGUUUUUGCCAUUACUUUCUAUGGCAAAAACUGUAAUUACUUUUGCACCAACCUAAUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008221
          miRBase ID: ptr-miR-548f
          Mature miRNA Sequence: AAAAACUGUAAUUACUUUU
          Chromosome Location: NA

          Stem-loop ID ptr-mir-548f-2
          Stem-loop Accession # MI0008751
          Stem-loop Sequence
          AUUAGGUUGGUGCAAACCUAAUUGCAAUUUUUGCAGUUUUUUUAAGUAAUUGCAAAAACUGUAAUUACUUUUGCACCAACCUAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008221
          miRBase ID: ptr-miR-548f
          Mature miRNA Sequence: AAAAACUGUAAUUACUUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305694_pri
          TaqMan™ Pri-miRNA Assay : Hs03305683_pri
          TaqMan™ Pri-miRNA Assay : Hs03305689_pri
          TaqMan™ Pri-miRNA Assay : Hs03305674_pri
          TaqMan™ Pri-miRNA Assay : Hs03305679_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13128
          Ambion® Pre-miR™ miRNA Precursor : PM13128
          mirVana® miRNA inhibitor : MH13128


          miRBase Alias:

          Human: hsa-miR-548f (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline