Search
Search

| Assay Name: | hsa-miR-548h-5p |
|---|---|
| miRBase Accession Number: | MI0006411 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAAGUAAUCGCGGUUUUUGUC |
| Species: | Human, Callithrix jacchus, Pan troglodytes |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUAAUCGCGGUUUUUGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548h-5p | MIMAT0005928 |
| Callithrix jacchus | cja-miR-548f | MIMAT0039576 |
| Pan troglodytes | ptr-miR-548h | MIMAT0008222 |