Search
Search

| Assay Name: | cin-miR-153-3p |
|---|---|
| miRBase Accession Number: | MI0007169 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCAUAGUAACAAAAGUGAUC |
| Species: | Ciona intestinalis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | UUGCAUAGUAACAAAAGUGAUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ciona intestinalis | cin-miR-153-3p | MIMAT0006104 |