Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13793
          Assay Name: hsa-miR-1302
          miRBase Accession Number: MI0006362
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 112695034 - 112695176 [-] on Build GRCh38
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Species: Human, Callithrix jacchus, Horse, Pan troglodytes, Pongo pygmaeus
          Product Type: mirVana® miRNA mimic
          Assay ID MC13793

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000456, mir-1302

          Mature miRNA Sequence:

          UUGGGACAUACUUAUGCUAAA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-1302 MIMAT0005890
          Callithrix jacchus cja-miR-1302 MIMAT0039634
          Horse eca-miR-1302d MIMAT0012969
          Pan troglodytes ptr-miR-1302 MIMAT0008021
          Pongo pygmaeus ppy-miR-1302 MIMAT0016222

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-1302-1
          Stem-loop Accession # MI0006362
          Stem-loop Sequence
          CAGAAAGCCCAGUUAAAUUUGAAUUUCAAGUAAACAAUGAAUAAUUGUGUAUGUAAGAAUAUCCCAUACAAUAUUUGGGACAUACUUAUGCUAAAAAUUAUUCCUUGCUUAUCUGAAAUUCAAAUGUAACUAGGAUUCCUGUA
          Chromosome Location Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-10
          Stem-loop Accession # MI0015979
          Stem-loop Sequence
          GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUUA
          Chromosome Location Chr. 15 - 101960459 - 101960596 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-11
          Stem-loop Accession # MI0015980
          Stem-loop Sequence
          GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUUA
          Chromosome Location Chr. 19 - 71973 - 72110 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-2
          Stem-loop Accession # MI0006363
          Stem-loop Sequence
          GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUUA
          Chromosome Location Chr. 1 - 30366 - 30503 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-3
          Stem-loop Accession # MI0006364
          Stem-loop Sequence
          GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUUUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUUA
          Chromosome Location Chr. 2 - 113582959 - 113583096 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-4
          Stem-loop Accession # MI0006365
          Stem-loop Sequence
          AAUGCAGAAGCACAGCUAAAAUUUGAAUUUCAGAUAAACAAAUUUUUCUUAGAAUAAGUAUGUCUCCAUGCAACAUUUGGGACAUACUUAUGCUAAAAUAUUAUUUGUGUUUCAUCUGAAAUUCAAAUUCAACUGGACAUCCUGUAUUUU
          Chromosome Location Chr. 2 - 207269275 - 207269424 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-5
          Stem-loop Accession # MI0006366
          Stem-loop Sequence
          UGCCCGGCCUCCCAUUAAAUUGGUUUUUCAGACAAAUCACAAAUUUGUUUAGGUAUAAGUAUAUCCCAUGUAAUCUUUGGGACAUACUUAUGCUAAAAUAAUUGUUCCUUGUUGAUUGGAAAUUUUAAUUUUAAUUAGGUGUCCUGUAUU
          Chromosome Location Chr. 20 - 50614636 - 50614785 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-6
          Stem-loop Accession # MI0006367
          Stem-loop Sequence
          AACAAAUAAUUUGGUAAUAUAUGUAUGGCCCACACAAUAUUUAGGACAACAAUAUUUGGGACAUACUUAUGCUAAAAAAGUAUUUGUUGA
          Chromosome Location Chr. 7 - 18127220 - 18127309 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-7
          Stem-loop Accession # MI0006368
          Stem-loop Sequence
          ACAACAUGUUUUUAGGACAUGUAUGUCUGGUGCAAUAAUUGGGACAUACUUAUGCUAAAAAAAUUAGUGUUC
          Chromosome Location Chr. 8 - 141786242 - 141786313 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-8
          Stem-loop Accession # MI0006369
          Stem-loop Sequence
          CCCAUUUAAACUUGAAUUUCAUAUAAACACCGUAAUUUUCAGCAUUAGUGUAUCACAUGCAGUAUUUGGGACAUACUUAUGCUAAAAAAUUAGGUGGUGUUGAUCUGAAAUUCCAGUGUAGAUGGGCA
          Chromosome Location Chr. 9 - 97363554 - 97363681 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1302-9
          Stem-loop Accession # MI0015978
          Stem-loop Sequence
          GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUUA
          Chromosome Location Chr. 9 - 30144 - 30281 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005890
          miRBase ID: hsa-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38

          Callithrix jacchus

          Stem-loop ID cja-mir-1302
          Stem-loop Accession # MI0032171
          Stem-loop Sequence
          AAAAUGCAGAAGCACAGCUAAAAUUUGAGUUUCAGAUAAACAAAUUUUUCUUAGAAUAAGUAUAUCUCCAUGCAACAUUUGGGACAUACUUAUGCUAAAACAUUAUUUGUGUUUCAUCUGAAAUUCAAAUUCAACUGGAUAUCCUGUAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039634
          miRBase ID: cja-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-1302d-4
          Stem-loop Accession # MI0012722
          Stem-loop Sequence
          UUGGAAUUUAGACAAAUAAUGAAGAAUUUUUUAGUGUAAGUAUGUCUUAUGCAAUAUUUGGGACAUACUUAUGCUAAAAAAUUUAUUAGUUAUCUGAAAUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012969
          miRBase ID: eca-miR-1302d
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-1302-1
          Stem-loop Accession # MI0008516
          Stem-loop Sequence
          CAGAAAGCCCAGUUAAAUUUGAAUUUCAAGUAAACAAUGAAUAAUUGUGUAUGUAAGAAUAUCCCAUACAAUAUUUGGGACAUACUUAUGCUAAAAAUUAUUCCUUGCUUAUCUGAAAUUCAAAUGUAACUAGGAUUCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008021
          miRBase ID: ptr-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1302-2
          Stem-loop Accession # MI0008517
          Stem-loop Sequence
          GGAUGCCCAGUUAGUUUGAAUUCUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008021
          miRBase ID: ptr-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1302-3
          Stem-loop Accession # MI0008518
          Stem-loop Sequence
          GAUGCCCAGUUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008021
          miRBase ID: ptr-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1302-4
          Stem-loop Accession # MI0008519
          Stem-loop Sequence
          GGAUGCCCAGUUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008021
          miRBase ID: ptr-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1302-5
          Stem-loop Accession # MI0008520
          Stem-loop Sequence
          GGAUGCCCAGUUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCUUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008021
          miRBase ID: ptr-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1302-6
          Stem-loop Accession # MI0008521
          Stem-loop Sequence
          AACAAAUAAUUUGGUAAUAUAUGUAUGGCCCACACAAUAUUUAGGACAACAAUAUUUGGGACAUACUUAUGCUAAAAAAGUAUUUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008021
          miRBase ID: ptr-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1302-7
          Stem-loop Accession # MI0008522
          Stem-loop Sequence
          ACAACAUGUUUUUAGUACAUGUAUGUCUGGUGCAAUAAUUGGGACAUACUUAUGCUAAAAAAAUUAGUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008021
          miRBase ID: ptr-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1302-8
          Stem-loop Accession # MI0008523
          Stem-loop Sequence
          CCCAUUUAAACUUGAAUUUCAUAUAAACACCGUAAUUUUCAGCAUUAGUGUAUCACAUGCAGUAUUUGGGACAUACUUAUGCUAAAAAAUUAGGUGGUGUUGAUCUGAAAUUCCAGUGUAGAUGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008021
          miRBase ID: ptr-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-1302-1
          Stem-loop Accession # MI0015256
          Stem-loop Sequence
          GGAUGCCCAGUUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCUUAGCCUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016222
          miRBase ID: ppy-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ppy-mir-1302-2
          Stem-loop Accession # MI0015260
          Stem-loop Sequence
          CCCAUUUAAACUUGAAUUUCAUAUAAACAGAGUAAUUUUCAGCAUUAGUGUAUCACAUGCAGUAUUUGGGACAUACUUAUGCUAAAAAAUUAGGUGGUGUUGAUCUGAAAUUCCAGUGUAGAUGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016222
          miRBase ID: ppy-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ppy-mir-1302-3
          Stem-loop Accession # MI0015258
          Stem-loop Sequence
          AACAAAUAAUUUGGUAAUAUAUGUAUGGCCCACACAAUAUUUAGGACAACAAUAUUUGGGACAUACUUAUGCUAAAAAAGUAUUUGUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016222
          miRBase ID: ppy-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA

          Stem-loop ID ppy-mir-1302-4
          Stem-loop Accession # MI0015259
          Stem-loop Sequence
          ACAACAUUUUUUUUAGUACAUGUAUGUCUGGUGCAACAAUUGGGACAUACUUAUGCUAAAAAAAUUAGUGUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016222
          miRBase ID: ppy-miR-1302
          Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305597_pri
          TaqMan™ Pri-miRNA Assay : Hs03305633_pri
          TaqMan™ Pri-miRNA Assay : Hs03305620_pri
          TaqMan™ Pri-miRNA Assay : Hs03305646_pri
          TaqMan™ Pri-miRNA Assay : Hs03305595_pri
          TaqMan™ Pri-miRNA Assay : Hs03305626_pri
          TaqMan™ Pri-miRNA Assay : Hs03305613_pri
          TaqMan™ Pri-miRNA Assay : Hs03305639_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13793
          Ambion® Pre-miR™ miRNA Precursor : PM13793
          TaqMan™ MicroRNA Assay : 002901
          mirVana® miRNA inhibitor : MH13793
          TaqMan™ Advanced miRNA Assay : 478697_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline