Search
Search

| Assay Name: | sja-let-7 |
|---|---|
| miRBase Accession Number: | MI0010668 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGAGGUAGUUCGUUGUGUGGU |
| Species: | Schistosoma japonicum, Schistosoma mansoni |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | GGAGGUAGUUCGUUGUGUGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schistosoma japonicum | sja-let-7 | MIMAT0010175 |
| Schistosoma mansoni | sma-let-7-5p | MIMAT0010180 |