Search
Search

| Assay Name: | lva-miR-153-3p |
|---|---|
| miRBase Accession Number: | MI0025154 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGAUU |
| Species: | Lytechinus variegatus, Patiria miniata, Saccoglossus kowalevskii, Strongylocentrotus purpuratus |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Lytechinus variegatus | lva-miR-153-3p | MIMAT0032208 |
| Patiria miniata | pmi-miR-153-3p | MIMAT0032161 |
| Saccoglossus kowalevskii | sko-miR-153 | MIMAT0009629 |
| Strongylocentrotus purpuratus | spu-miR-153-3p | MIMAT0009669 |