Search
Search

| Assay Name: | dme-miR-2279-3p |
|---|---|
| miRBase Accession Number: | MI0011289 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUCACGCGAAGAUAUUUAUUU |
| Species: | Drosophila melanogaster |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UUUCACGCGAAGAUAUUUAUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-2279-3p | MIMAT0011785 |