Search
Search

| Assay Name: | hvt-miR-H12-5p |
|---|---|
| miRBase Accession Number: | MI0012625 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUCUUAUGGACCGCCGAGGUGUGUC |
| Species: | Herpesvirus of turkeys |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000783, hvt-mir-H9 |
Mature miRNA Sequence: | UUUCUUAUGGACCGCCGAGGUGUGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Herpesvirus of turkeys | hvt-miR-H12-5p | MIMAT0012863 |