Search
Search

| Assay Name: | dps-miR-2526-3p |
|---|---|
| miRBase Accession Number: | MI0011659 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCCGUCGGCGUGUUGCGGUG |
| Species: | Drosophila pseudoobscura |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | GCCGUCGGCGUGUUGCGGUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila pseudoobscura | dps-miR-2526-3p | MIMAT0012315 |