Search
Search

| Assay Name: | cte-miR-1990c-3p |
|---|---|
| miRBase Accession Number: | MI0012130 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGGACUAUGUCAACUUACAAC |
| Species: | Capitella teleta |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000718, mir-1990 |
Mature miRNA Sequence: | UGGGACUAUGUCAACUUACAAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Capitella teleta | cte-miR-1990c-3p | MIMAT0013548 |