Search
Search

| Assay Name: | bhv1-miR-B8-3p |
|---|---|
| miRBase Accession Number: | MI0013180 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUUGUAUUUUUGCGCCAGCGGG |
| Species: | Bovine herpesvirus 1 |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0002021, mir-B8 |
Mature miRNA Sequence: | UAUUGUAUUUUUGCGCCAGCGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine herpesvirus 1 | bhv1-miR-B8-3p | MIMAT0013971 |