Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19255
          Assay Name: mmu-miR-1a-2-5p
          miRBase Accession Number: MI0000652
          miRBase Version: v22.1
          Chromosome Location: Chr.18: 10785481 - 10785552 [-] on Build GRCm38
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Species: Mouse, Alligator mississippiensis, Chicken, Chrysemys picta, Columba livia, Dasypus novemcinctus, Guinea pig, Monodelphis domestica, Ophiophagus hannah, Ornithorhynchus anatinus, Pteropus alecto, Rabbit, Rhesus monkey, Xenopus laevis
          Product Type: mirVana® miRNA mimic
          Assay ID MC19255

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000038, mir-1

          Mature miRNA Sequence:

          ACAUACUUCUUUAUGUACCCAUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-1a-2-5p MIMAT0017047
          Alligator mississippiensis ami-miR-1a-2-5p MIMAT0038020
          Chicken gga-miR-1a-2-5p MIMAT0026510
          Chrysemys picta cpi-miR-1a-2-5p MIMAT0037614
          Columba livia cli-miR-1a-2-5p MIMAT0038393
          Dasypus novemcinctus dno-miR-1-2-5p MIMAT0047490
          View More Rows
          Guinea pig cpo-miR-1a-5p MIMAT0046837
          Monodelphis domestica mdo-miR-1-1-5p MIMAT0026647
          Ophiophagus hannah oha-miR-1a-5p MIMAT0036646
          Ornithorhynchus anatinus oan-miR-1a-5p MIMAT0026752
          Pteropus alecto pal-miR-1-5p MIMAT0039767
          Rabbit ocu-miR-1-5p MIMAT0048091
          Rhesus monkey mml-miR-1-2-5p MIMAT0031099
          Xenopus laevis xla-miR-1a-5p MIMAT0046516

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-1a-2
          Stem-loop Accession # MI0000652
          Stem-loop Sequence
          UCAGAGCACAUACUUCUUUAUGUACCCAUAUGAACAUUCAGUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUG
          Chromosome Location Chr. 18 - 10785481 - 10785552 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017047
          miRBase ID: mmu-miR-1a-2-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: Chr. 18 - 10785481 - 10785552 [-] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-1a-2
          Stem-loop Accession # MI0029653
          Stem-loop Sequence
          ACCUGCCCAGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGCAGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038020
          miRBase ID: ami-miR-1a-2-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-1a-2
          Stem-loop Accession # MI0001196
          Stem-loop Sequence
          ACCUGCUCAGAGCACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026510
          miRBase ID: gga-miR-1a-2-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-1a-2
          Stem-loop Accession # MI0029385
          Stem-loop Sequence
          UCCUGCCCAGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUAUAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGCAGGCACUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037614
          miRBase ID: cpi-miR-1a-2-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-1a-2
          Stem-loop Accession # MI0029894
          Stem-loop Sequence
          GUACCUGCCCAGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGCAGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038393
          miRBase ID: cli-miR-1a-2-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-1-2
          Stem-loop Accession # MI0038896
          Stem-loop Sequence
          ACAUACUUCUUUAUGUACCCAUAUGAACAUAUAAUGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047490
          miRBase ID: dno-miR-1-2-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-1a
          Stem-loop Accession # MI0038552
          Stem-loop Sequence
          ACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046837
          miRBase ID: cpo-miR-1a-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-1-1
          Stem-loop Accession # MI0005269
          Stem-loop Sequence
          ACCUACUCAGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGUAGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026647
          miRBase ID: mdo-miR-1-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-1a
          Stem-loop Accession # MI0031312
          Stem-loop Sequence
          GAAAACCUGCCCAGAGCACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUAUUAUGGAAUGUAAAGAAGUAUGUACUUUUGGCAGGAGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036646
          miRBase ID: oha-miR-1a-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-1a
          Stem-loop Accession # MI0006700
          Stem-loop Sequence
          GGGAUGUACACCUGCUCAGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUAAAAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGCAGGCGCCGACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026752
          miRBase ID: oan-miR-1a-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-1-1
          Stem-loop Accession # MI0032317
          Stem-loop Sequence
          ACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039767
          miRBase ID: pal-miR-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-1-2
          Stem-loop Accession # MI0039215
          Stem-loop Sequence
          ACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048091
          miRBase ID: ocu-miR-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-1-2
          Stem-loop Accession # MI0007569
          Stem-loop Sequence
          CAGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031099
          miRBase ID: mml-miR-1-2-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-1a
          Stem-loop Accession # MI0038286
          Stem-loop Sequence
          ACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046516
          miRBase ID: xla-miR-1a-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUGUACCCAUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03308741_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19255
          Ambion® Pre-miR™ miRNA Precursor : PM19255
          TaqMan™ MicroRNA Assay : 464216_mat
          mirVana® miRNA inhibitor : MH19255


          miRBase Alias:

          Mouse: mmu-miR-1a-2* (v17)
          Mouse: mmu-miR-1-2* (v16)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline