Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19454
          Assay Name: mmu-miR-1a-1-5p
          miRBase Accession Number: MI0000139
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 180389048 - 180389124 [+] on Build GRCm38
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Species: Mouse, Chicken, Chrysemys picta, Columba livia, Dasypus novemcinctus, Ictalurus punctatus, Ophiophagus hannah, Paralichthys olivaceus, Rhesus monkey, Salmo salar, Taeniopygia guttata
          Product Type: mirVana® miRNA mimic
          Assay ID MC19454

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000038, mir-1

          Mature miRNA Sequence:

          ACAUACUUCUUUAUAUGCCCAUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-1a-1-5p MIMAT0016979
          Chicken gga-miR-1a-1-5p MIMAT0031071
          Chrysemys picta cpi-miR-1a-5p MIMAT0037612
          Columba livia cli-miR-1a-5p MIMAT0038391
          Dasypus novemcinctus dno-miR-1-5p MIMAT0047488
          Ictalurus punctatus ipu-miR-1 MIMAT0029384
          View More Rows
          Ophiophagus hannah oha-miR-1b-5p MIMAT0036799
          Paralichthys olivaceus pol-miR-1-5p MIMAT0025412
          Rhesus monkey mml-miR-1-1-5p MIMAT0026788
          Salmo salar ssa-miR-1-4-5p MIMAT0032474
          Taeniopygia guttata tgu-miR-1-1-5p MIMAT0031087

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-1a-1
          Stem-loop Accession # MI0000139
          Stem-loop Sequence
          GCUUGGGACACAUACUUCUUUAUAUGCCCAUAUGAACCUGCUAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGC
          Chromosome Location Chr. 2 - 180389048 - 180389124 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0016979
          miRBase ID: mmu-miR-1a-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: Chr. 2 - 180389048 - 180389124 [+] on Build GRCm38

          Chicken

          Stem-loop ID gga-mir-1a-1
          Stem-loop Accession # MI0001247
          Stem-loop Sequence
          UGAGAGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCAAUCUAUGGAAUGUAAAGAAGUAUGUAUUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031071
          miRBase ID: gga-miR-1a-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-1a-1
          Stem-loop Accession # MI0029384
          Stem-loop Sequence
          CUACCUGCUUGAGAGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGUGGGGAUCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037612
          miRBase ID: cpi-miR-1a-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-1a-1
          Stem-loop Accession # MI0029893
          Stem-loop Sequence
          CUGCUUGAGAGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGGAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038391
          miRBase ID: cli-miR-1a-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-1-1
          Stem-loop Accession # MI0038895
          Stem-loop Sequence
          ACAUACUUCUUUAUAUGCCCAUAUGGAGCUCGGAGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047488
          miRBase ID: dno-miR-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-1-1
          Stem-loop Accession # MI0024455
          Stem-loop Sequence
          ACAUACUUCUUUAUAUGCCCAUAUGAACAAGAGCAGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029384
          miRBase ID: ipu-miR-1
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-1b
          Stem-loop Accession # MI0031397
          Stem-loop Sequence
          AAUGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCCAACUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGCACGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036799
          miRBase ID: oha-miR-1b-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-1
          Stem-loop Accession # MI0022189
          Stem-loop Sequence
          ACAUACUUCUUUAUAUGCCCAUAUGAACAAGAGCAACUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025412
          miRBase ID: pol-miR-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-1-1
          Stem-loop Accession # MI0007568
          Stem-loop Sequence
          GCUUGGGAAACAUACUUCUUUAUAUGCCCAUAUGGACCUGCUAAGCUAUGGAAUGUAAAGAAGUAUGUAUCUCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026788
          miRBase ID: mml-miR-1-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-1-4
          Stem-loop Accession # MI0026580
          Stem-loop Sequence
          ACAUACUUCUUUAUAUGCCCAUAUGAACAAGAGCAACUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032474
          miRBase ID: ssa-miR-1-4-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-1-1
          Stem-loop Accession # MI0013781
          Stem-loop Sequence
          GACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031087
          miRBase ID: tgu-miR-1-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306163_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19454
          Ambion® Pre-miR™ miRNA Precursor : PM19454
          TaqMan™ MicroRNA Assay : 462049_mat
          mirVana® miRNA inhibitor : MH19454


          miRBase Alias:

          Mouse: mmu-miR-1a-1* (v17)
          Mouse: mmu-miR-1-1* (v16)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline