Search
Search

| Assay Name: | ssc-miR-155-3p |
|---|---|
| miRBase Accession Number: | MI0015907 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCUACAUGUUAGCAUUAACA |
| Species: | Pig |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000157, mir-155 |
Mature miRNA Sequence: | UCCUACAUGUUAGCAUUAACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-155-3p | MIMAT0017953 |