Search
Search

| Assay Name: | mmu-miR-743a-5p |
|---|---|
| miRBase Accession Number: | MI0005207 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUUCAGAUUGGUGCCUGUCAU |
| Species: | Mouse, Rat |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UAUUCAGAUUGGUGCCUGUCAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-743a-5p | MIMAT0017263 |
| Rat | rno-miR-743a-5p | MIMAT0017315 |