Search
Search

| Assay Name: | mmu-miR-3077-3p |
|---|---|
| miRBase Accession Number: | MI0014040 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGACUCCCUGCUUCUCCGCAG |
| Species: | Mouse |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | CUGACUCCCUGCUUCUCCGCAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3077-3p | MIMAT0014863 |