Search
Search

| Assay Name: | pma-miR-194-5p |
|---|---|
| miRBase Accession Number: | MI0017098 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUAACAGCAACUCCAUGUGGAU |
| Species: | Petromyzon marinus |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000055, mir-194 |
Mature miRNA Sequence: | UGUAACAGCAACUCCAUGUGGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Petromyzon marinus | pma-miR-194-5p | MIMAT0019500 |