Search
Search

| Assay Name: | hsa-miR-4444 |
|---|---|
| miRBase Accession Number: | MI0016787 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCGAGUUGGAAGAGGCG |
| Species: | Human, Bovine |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0001310, mir-4444 |
Mature miRNA Sequence: | CUCGAGUUGGAAGAGGCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4444 | MIMAT0018962 |
| Bovine | bta-miR-4444 | MIMAT0036978 |