Search
Search

| Assay Name: | rlcv-miR-rL1-28-5p |
|---|---|
| miRBase Accession Number: | MI0013287 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAAGGCGUUGCUUUCAGCUG |
| Species: | Rhesus lymphocryptovirus |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UGGAAGGCGUUGCUUUCAGCUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rhesus lymphocryptovirus | rlcv-miR-rL1-28-5p | MIMAT0019193 |