Search
Search

| Assay Name: | pma-miR-29a-3p |
|---|---|
| miRBase Accession Number: | MI0017035 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGCACCAUUUGAAAUCAGUUA |
| Species: | Petromyzon marinus, Salmo salar |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUUUGAAAUCAGUUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Petromyzon marinus | pma-miR-29a-3p | MIMAT0019417 |
| Salmo salar | ssa-miR-29c-3p | MIMAT0032591 |