Search
Search

| Assay Name: | ath-miR5020a |
|---|---|
| miRBase Accession Number: | MI0017888 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAAGAAGGUGAGACUUGCA |
| Species: | Arabidopsis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0001226, MIR5020 |
Mature miRNA Sequence: | UGGAAGAAGGUGAGACUUGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR5020a | MIMAT0020524 |