Search
Search

| Assay Name: | aca-miR-5400 |
|---|---|
| miRBase Accession Number: | MI0018902 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAGCACUCUGCAGGAAAGAAC |
| Species: | Anolis carolinensis |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UUAGCACUCUGCAGGAAAGAAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-5400 | MIMAT0022010 |