Search
Search

| Assay Name: | ame-miR-6002-5p |
|---|---|
| miRBase Accession Number: | MI0020212 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCGAAGCGAAGGGUAUUCC |
| Species: | Apis mellifera |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UCGAAGCGAAGGGUAUUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Apis mellifera | ame-miR-6002-5p | MIMAT0023543 |