Search
Search

| Assay Name: | mmu-miR-6985-3p |
|---|---|
| miRBase Accession Number: | MI0022833 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCACAGCCACCUCCUCACAG |
| Species: | Mouse |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | CUCACAGCCACCUCCUCACAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-6985-3p | MIMAT0027873 |