Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC28440
          Assay Name: hsa-miR-155-5p
          miRBase Accession Number: MI0000681
          miRBase Version: v22.1
          Chromosome Location: Chr.21: 25573980 - 25574044 [+] on Build GRCh38
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Species: Human, Dasypus novemcinctus, Guinea pig, Ictalurus punctatus, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Rabbit
          Product Type: mirVana® miRNA mimic
          Assay ID MC28440

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000157, mir-155

          Mature miRNA Sequence:

          UUAAUGCUAAUCGUGAUAGGGGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-155-5p MIMAT0000646
          Dasypus novemcinctus dno-miR-155-5p MIMAT0047687
          Guinea pig cpo-miR-155-5p MIMAT0047038
          Ictalurus punctatus ipu-miR-155 MIMAT0029429
          Nomascus leucogenys nle-miR-155 MIMAT0049433
          Otolemur garnettii oga-miR-155 MIMAT0049607
          View More Rows
          Pan paniscus ppa-miR-155 MIMAT0049073
          Rabbit ocu-miR-155-5p MIMAT0048273

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-155
          Stem-loop Accession # MI0000681
          Stem-loop Sequence
          CUGUUAAUGCUAAUCGUGAUAGGGGUUUUUGCCUCCAACUGACUCCUACAUAUUAGCAUUAACAG
          Chromosome Location Chr. 21 - 25573980 - 25574044 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000646
          miRBase ID: hsa-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Chromosome Location: Chr. 21 - 25573980 - 25574044 [+] on Build GRCh38

          Dasypus novemcinctus

          Stem-loop ID dno-mir-155
          Stem-loop Accession # MI0039006
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCCAACUAACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047687
          miRBase ID: dno-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-155
          Stem-loop Accession # MI0038664
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCUGACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047038
          miRBase ID: cpo-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-155
          Stem-loop Accession # MI0024518
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUAGUGCUGAUGAACACCUAUGCUGUUAGCAUUAAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029429
          miRBase ID: ipu-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-155
          Stem-loop Accession # MI0040178
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCCAACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049433
          miRBase ID: nle-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-155
          Stem-loop Accession # MI0040388
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCUGACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049607
          miRBase ID: oga-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-155
          Stem-loop Accession # MI0039781
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUGCCUCCAACUGACUCCUACAUAUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049073
          miRBase ID: ppa-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-155
          Stem-loop Accession # MI0039314
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCCAACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048273
          miRBase ID: ocu-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303349_pri
          TaqMan™ MicroRNA Assay : 467534_mat
          mirVana® miRNA inhibitor : MH28440
          TaqMan™ Advanced miRNA Assay : 483064_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline