Search
Search

| Assay Name: | lja-miR7518 |
|---|---|
| miRBase Accession Number: | MI0024392 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCGCACUGAGCAAGGACAGG |
| Species: | Lotus japonicus |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UUGCGCACUGAGCAAGGACAGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Lotus japonicus | lja-miR7518 | MIMAT0029333 |