Search
Search

| Assay Name: | hsa-miR-598-5p |
|---|---|
| miRBase Accession Number: | MI0003610 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCGGUGAUCCCGAUGGUGUGAGC |
| Species: | Human, Rhesus monkey |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000393, mir-598 |
Mature miRNA Sequence: | GCGGUGAUCCCGAUGGUGUGAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-598-5p | MIMAT0026620 |
| Rhesus monkey | mml-miR-598-5p | MIMAT0026902 |