Search
Search

| Assay Name: | tch-miR-361-5p |
|---|---|
| miRBase Accession Number: | MI0031175 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAUCAGAAUCUCCAGGGGUA |
| Species: | Tupaia chinensis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000172, mir-361 |
Mature miRNA Sequence: | UUAUCAGAAUCUCCAGGGGUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tupaia chinensis | tch-miR-361-5p | MIMAT0036517 |