Search
Search

| Assay Name: | sma-miR-8455-5p |
|---|---|
| miRBase Accession Number: | MI0027252 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUUUGGUUAACAUUCUACC |
| Species: | Schistosoma mansoni |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | AUUUGGUUAACAUUCUACC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schistosoma mansoni | sma-miR-8455-5p | MIMAT0033622 |