Search
Search

| Assay Name: | sfr-miR-10464-5p |
|---|---|
| miRBase Accession Number: | MI0033558 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACACGACUGCAAUAGAACUG |
| Species: | Spodoptera frugiperda |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | ACACGACUGCAAUAGAACUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Spodoptera frugiperda | sfr-miR-10464-5p | MIMAT0041798 |