Search
Search

| Assay Name: | ocu-miR-28-3p |
|---|---|
| miRBase Accession Number: | MI0039242 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CACUAGAUUAUGAGCUCCUGGA |
| Species: | Otolemur garnettii, Rabbit |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | CACUAGAUUAUGAGCUCCUGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Otolemur garnettii | oga-miR-28a | MIMAT0049621 |
| Rabbit | ocu-miR-28-3p | MIMAT0048142 |