Search
Search

| Assay Name: | pbv-miR-29a-3p |
|---|---|
| miRBase Accession Number: | MI0030179 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAGCACCAUUUGAAAUCGGU |
| Species: | Python bivittatus |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | CUAGCACCAUUUGAAAUCGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Python bivittatus | pbv-miR-29a-3p | MIMAT0038866 |