Search
Search

| Assay Name: | ath-miR169b-5p |
|---|---|
| miRBase Accession Number: | MI0000976 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGCCAAGGAUGACUUGCCGG |
| Species: | Arabidopsis, Aegilops tauschii, Aquilegia caerulea, Arabidopsis lyrata, Brachypodium distachyon, Brassica napus, Camelina sativa, Citrus sinensis, Cucumis melo, Glycine max, Linum usitatissimum, Malus domestica, Manihot esculenta, Medicago truncatula, Nicotiana tabacum, Populus trichocarpa, Prunus persica, Rice, Ricinus communis, Solanum lycopersicum, Sorghum bicolor, Theobroma cacao, Vigna unguiculata, Vitis vinifera, Zea mays |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000037, MIR169_2 |
Mature miRNA Sequence: | CAGCCAAGGAUGACUUGCCGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR169b-5p | MIMAT0000906 |
| Arabidopsis | ath-miR169c | MIMAT0000907 |
| Aegilops tauschii | ata-miR169d-5p | MIMAT0037206 |
| Aquilegia caerulea | aqc-miR169c | MIMAT0012562 |
| Arabidopsis lyrata | aly-miR169b-5p | MIMAT0017487 |
| Arabidopsis lyrata | aly-miR169c-5p | MIMAT0017489 |
| View More Rows | ||
| Brachypodium distachyon | bdi-miR169c-5p | MIMAT0020701 |
| Brachypodium distachyon | bdi-miR169f | MIMAT0020736 |
| Brassica napus | bna-miR169n | MIMAT0023629 |
| Camelina sativa | cas-miR169b | MIMAT0045311 |
| Citrus sinensis | csi-miR169g-5p | MIMAT0048896 |
| Citrus sinensis | csi-miR169k-5p | MIMAT0048900 |
| Citrus sinensis | csi-miR169m-5p | MIMAT0048903 |
| Citrus sinensis | csi-miR169n-5p | MIMAT0048904 |
| Citrus sinensis | csi-miR169o-5p | MIMAT0048995 |
| Citrus sinensis | csi-miR169r-5p | MIMAT0049005 |
| Cucumis melo | cme-miR169f | MIMAT0026115 |
| Cucumis melo | cme-miR169h | MIMAT0026114 |
| Glycine max | gma-miR169a | MIMAT0001693 |
| Glycine max | gma-miR169f | MIMAT0020986 |
| Glycine max | gma-miR169g | MIMAT0020987 |
| Glycine max | gma-miR169m | MIMAT0021653 |
| Linum usitatissimum | lus-miR169f | MIMAT0027148 |
| Linum usitatissimum | lus-miR169h | MIMAT0027198 |
| Linum usitatissimum | lus-miR169j | MIMAT0027200 |
| Malus domestica | mdm-miR169a | MIMAT0025934 |
| Malus domestica | mdm-miR169g | MIMAT0043575 |
| Malus domestica | mdm-miR169h | MIMAT0043576 |
| Malus domestica | mdm-miR169i | MIMAT0043577 |
| Malus domestica | mdm-miR169j | MIMAT0043578 |
| Manihot esculenta | mes-miR169a | MIMAT0029209 |
| Manihot esculenta | mes-miR169b | MIMAT0029210 |
| Manihot esculenta | mes-miR169c | MIMAT0029211 |
| Manihot esculenta | mes-miR169d | MIMAT0029212 |
| Manihot esculenta | mes-miR169e | MIMAT0029213 |
| Medicago truncatula | mtr-miR169b | MIMAT0011111 |
| Medicago truncatula | mtr-miR169g | MIMAT0011090 |
| Medicago truncatula | mtr-miR169l-5p | MIMAT0030001 |
| Nicotiana tabacum | nta-miR169q | MIMAT0024682 |
| Nicotiana tabacum | nta-miR169r | MIMAT0024683 |
| Nicotiana tabacum | nta-miR169s | MIMAT0024684 |
| Populus trichocarpa | ptc-miR169d | MIMAT0001960 |
| Populus trichocarpa | ptc-miR169e | MIMAT0001961 |
| Populus trichocarpa | ptc-miR169f | MIMAT0001962 |
| Populus trichocarpa | ptc-miR169g | MIMAT0001963 |
| Populus trichocarpa | ptc-miR169h | MIMAT0001964 |
| Populus trichocarpa | ptc-miR169p | MIMAT0001972 |
| Prunus persica | ppe-miR169a | MIMAT0031455 |
| Prunus persica | ppe-miR169b | MIMAT0031456 |
| Prunus persica | ppe-miR169c | MIMAT0031457 |
| Rice | osa-miR169b | MIMAT0001047 |
| Rice | osa-miR169c | MIMAT0001048 |
| Ricinus communis | rco-miR169a | MIMAT0014169 |
| Ricinus communis | rco-miR169b | MIMAT0014170 |
| Solanum lycopersicum | sly-miR169a | MIMAT0007918 |
| Sorghum bicolor | sbi-miR169b | MIMAT0001409 |
| Sorghum bicolor | sbi-miR169k | MIMAT0011336 |
| Theobroma cacao | tcc-miR169b | MIMAT0020392 |
| Theobroma cacao | tcc-miR169k | MIMAT0020401 |
| Theobroma cacao | tcc-miR169l | MIMAT0020402 |
| Vigna unguiculata | vun-miR169 | MIMAT0022759 |
| Vitis vinifera | vvi-miR169a | MIMAT0005676 |
| Vitis vinifera | vvi-miR169c | MIMAT0005678 |
| Vitis vinifera | vvi-miR169j | MIMAT0005683 |
| Vitis vinifera | vvi-miR169k | MIMAT0005684 |
| Vitis vinifera | vvi-miR169s | MIMAT0005688 |
| Vitis vinifera | vvi-miR169w | MIMAT0006553 |
| Zea mays | zma-miR169c-5p | MIMAT0001728 |
| Zea mays | zma-miR169r-5p | MIMAT0013994 |