Search
Search

| Assay Name: | ath-miR169a-5p |
|---|---|
| miRBase Accession Number: | MI0000212 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGCCAAGGAUGACUUGCCGA |
| Species: | Arabidopsis, Aegilops tauschii, Arabidopsis lyrata, Brachypodium distachyon, Brassica napus, Camelina sativa, Glycine max, Linum usitatissimum, Manihot esculenta, Medicago truncatula, Nicotiana tabacum, Populus trichocarpa, Rice, Solanum lycopersicum, Sorghum bicolor, Theobroma cacao, Vitis vinifera, Zea mays |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000037, MIR169_2 |
Mature miRNA Sequence: | CAGCCAAGGAUGACUUGCCGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR169a-5p | MIMAT0000200 |
| Aegilops tauschii | ata-miR169e-5p | MIMAT0037115 |
| Aegilops tauschii | ata-miR169f-5p | MIMAT0037178 |
| Aegilops tauschii | ata-miR169g-5p | MIMAT0037086 |
| Arabidopsis lyrata | aly-miR169a-5p | MIMAT0017485 |
| Brachypodium distachyon | bdi-miR169a-5p | MIMAT0020695 |
| View More Rows | ||
| Brassica napus | bna-miR169a | MIMAT0005612 |
| Brassica napus | bna-miR169b | MIMAT0005613 |
| Camelina sativa | cas-miR169a | MIMAT0045310 |
| Glycine max | gma-miR169b | MIMAT0007356 |
| Linum usitatissimum | lus-miR169g | MIMAT0027196 |
| Linum usitatissimum | lus-miR169l | MIMAT0027215 |
| Manihot esculenta | mes-miR169g | MIMAT0029215 |
| Medicago truncatula | mtr-miR169a | MIMAT0001643 |
| Nicotiana tabacum | nta-miR169a | MIMAT0024667 |
| Nicotiana tabacum | nta-miR169b | MIMAT0024668 |
| Nicotiana tabacum | nta-miR169c | MIMAT0024669 |
| Nicotiana tabacum | nta-miR169d | MIMAT0024670 |
| Nicotiana tabacum | nta-miR169e | MIMAT0024671 |
| Nicotiana tabacum | nta-miR169f | MIMAT0024672 |
| Nicotiana tabacum | nta-miR169g | MIMAT0024673 |
| Nicotiana tabacum | nta-miR169h | MIMAT0024674 |
| Nicotiana tabacum | nta-miR169i | MIMAT0024675 |
| Nicotiana tabacum | nta-miR169j | MIMAT0024676 |
| Nicotiana tabacum | nta-miR169k | MIMAT0024677 |
| Nicotiana tabacum | nta-miR169l | MIMAT0024678 |
| Nicotiana tabacum | nta-miR169m | MIMAT0024679 |
| Nicotiana tabacum | nta-miR169o | MIMAT0024680 |
| Nicotiana tabacum | nta-miR169p | MIMAT0024681 |
| Populus trichocarpa | ptc-miR169a | MIMAT0001951 |
| Populus trichocarpa | ptc-miR169b-5p | MIMAT0001958 |
| Populus trichocarpa | ptc-miR169c | MIMAT0001959 |
| Rice | osa-miR169a | MIMAT0000644 |
| Solanum lycopersicum | sly-miR169c | MIMAT0007920 |
| Sorghum bicolor | sbi-miR169a | MIMAT0001410 |
| Theobroma cacao | tcc-miR169a | MIMAT0020391 |
| Theobroma cacao | tcc-miR169c | MIMAT0020393 |
| Theobroma cacao | tcc-miR169e | MIMAT0020395 |
| Vitis vinifera | vvi-miR169f | MIMAT0005681 |
| Vitis vinifera | vvi-miR169g | MIMAT0005682 |
| Zea mays | zma-miR169a-5p | MIMAT0001368 |
| Zea mays | zma-miR169b-5p | MIMAT0001369 |