Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH11124
          Assay Name: dme-miR-263b-5p
          miRBase Accession Number: MI0000383
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Species: Drosophila melanogaster, Anopheles gambiae, Bombyx mori, Drosophila ananassae, Drosophila erecta, Drosophila grimshawi, Drosophila persimilis, Drosophila pseudoobscura, Drosophila sechellia, Drosophila simulans, Drosophila willistoni, Drosophila yakuba
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH11124

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000122, mir-263

          Mature miRNA Sequence:

          CUUGGCACUGGGAGAAUUCAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Drosophila melanogaster dme-miR-263b-5p MIMAT0000362
          Anopheles gambiae aga-miR-263b MIMAT0010111
          Bombyx mori bmo-miR-263b-5p MIMAT0004199
          Drosophila ananassae dan-miR-263b MIMAT0008447
          Drosophila erecta der-miR-263b MIMAT0008516
          Drosophila grimshawi dgr-miR-263b MIMAT0008564
          View More Rows
          Drosophila persimilis dpe-miR-263b MIMAT0008735
          Drosophila pseudoobscura dps-miR-263b MIMAT0001236
          Drosophila sechellia dse-miR-263b MIMAT0008830
          Drosophila simulans dsi-miR-263b MIMAT0008871
          Drosophila willistoni dwi-miR-263b MIMAT0009002
          Drosophila yakuba dya-miR-263b MIMAT0009110

          Stem-loop Details



          Drosophila melanogaster

          Stem-loop ID dme-mir-263b
          Stem-loop Accession # MI0000383
          Stem-loop Sequence
          UUGCUGACUUUGAGUCUUGGCACUGGGAGAAUUCACAGUUGACUUUAUUAUUCUGUGGUUCUGCGGGUGCCAAAACUUAAAAACCGGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000362
          miRBase ID: dme-miR-263b-5p
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Anopheles gambiae

          Stem-loop ID aga-mir-263b
          Stem-loop Accession # MI0010606
          Stem-loop Sequence
          UGACCAAUUAUGGACCUUGGCACUGGGAGAAUUCACAGUGGAUCGUACCAUAUCGUUUCUGUGGAUCUUUUCGUGCCAUCGUUCAGAUUUGGUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010111
          miRBase ID: aga-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Bombyx mori

          Stem-loop ID bmo-mir-263b
          Stem-loop Accession # MI0004977
          Stem-loop Sequence
          AGCCGACGCUGUUCCUUGGCACUGGGAGAAUUCACAGGAGUUGUAAUUCAUACCCGUGAAUUUCCCGAUGCCUUAGCUCAGUGUGGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004199
          miRBase ID: bmo-miR-263b-5p
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila ananassae

          Stem-loop ID dan-mir-263b
          Stem-loop Accession # MI0008981
          Stem-loop Sequence
          UGCUGUCUUUGAAUCUUGGCACUGGGAGAAUUCACAGUUGAUUUUAUUAUUCUGUGGUUCUGCGGGUGCCAAAAUUGAAAUAAUGGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008447
          miRBase ID: dan-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila erecta

          Stem-loop ID der-mir-263b
          Stem-loop Accession # MI0009055
          Stem-loop Sequence
          UAGCUGACUUUGAGUCUUGGCACUGGGAGAAUUCACAGUUGAUUUUAUUAUUCUGUGGUUCUGCGGGUGCCAAAAUUUAAUCAACAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008516
          miRBase ID: der-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila grimshawi

          Stem-loop ID dgr-mir-263b
          Stem-loop Accession # MI0009107
          Stem-loop Sequence
          CAUUACAUCUUGGCACUGGGAGAAUUCACAGUUGGUUAUAUAAUAUUCUGUGGUUCUGCGGGUGCCAAACUGUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008564
          miRBase ID: dgr-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila persimilis

          Stem-loop ID dpe-mir-263b
          Stem-loop Accession # MI0009289
          Stem-loop Sequence
          UUGCUGGCAUUGGCUCUUGGCACUGGGAGAAUUCACAGUUGGUUUUCAUAUAUUCUGUGGUUCUGCGGGUGCCCAAAAUCGAAUGUUCGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008735
          miRBase ID: dpe-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila pseudoobscura

          Stem-loop ID dps-mir-263b
          Stem-loop Accession # MI0001330
          Stem-loop Sequence
          UUGCUGGCAUUGGCUCUUGGCACUGGGAGAAUUCACAGUUGGUUUCCAUAUAUUCUGUGGUUCUGCGGGUGCCAAAAUCGAAUGUUCGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001236
          miRBase ID: dps-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila sechellia

          Stem-loop ID dse-mir-263b
          Stem-loop Accession # MI0009393
          Stem-loop Sequence
          UUGCUGACUUUGAGUCUUGGCACUGGGAGAAUUCACAGUUGACUUGAUUAUUCUGUGGUUCUGCGGGUGCCAAAACUUAAAAACCGGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008830
          miRBase ID: dse-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila simulans

          Stem-loop ID dsi-mir-263b
          Stem-loop Accession # MI0009436
          Stem-loop Sequence
          UUGCUGACUUUGAGUCUUGGCACUGGGAGAAUUCACAGUUGACUUGAUUAUUCUGUGGUUCUGCGGGUGCCAAAACUUAAAAACCGGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008871
          miRBase ID: dsi-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila willistoni

          Stem-loop ID dwi-mir-263b
          Stem-loop Accession # MI0009568
          Stem-loop Sequence
          AUCUUGGCACUGGGAGAAUUCACAGUUGAUUCUUAUUCGAUAUUCUGUGGUUCUGCGGGUGCCAAAAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009002
          miRBase ID: dwi-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA

          Drosophila yakuba

          Stem-loop ID dya-mir-263b
          Stem-loop Accession # MI0009687
          Stem-loop Sequence
          UGCUGACUUUGAGUCUUGGCACUGGGAGAAUUCACAGUUGACUUUAUUAUUCUGUGGUUCUGCGGGUGCCAAAAUUUAAUCAACGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009110
          miRBase ID: dya-miR-263b
          Mature miRNA Sequence: CUUGGCACUGGGAGAAUUCAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 000294
          mirVana® miRNA mimic : MC11124


          miRBase Alias:

          Drosophila melanogaster: dme-miR-263b (v16)
          Bombyx mori: bmo-miR-263b (v18)
          Drosophila virilis: dvi-miR-263b (v20)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline