Search
Search

| Assay Name: | ebv-miR-BART5-5p |
|---|---|
| miRBase Accession Number: | MI0003727 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAGGUGAAUAUAGCUGCCCAUCG |
| Species: | Epstein Barr virus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000326, mir-BART5 |
Mature miRNA Sequence: | CAAGGUGAAUAUAGCUGCCCAUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Epstein Barr virus | ebv-miR-BART5-5p | MIMAT0003413 |