Search
Search

| Assay Name: | ath-miR161.1 |
|---|---|
| miRBase Accession Number: | MI0000193 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAAAGUGACUACAUCGGGGU |
| Species: | Arabidopsis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000455, MIR161 |
Mature miRNA Sequence: | UGAAAGUGACUACAUCGGGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR161.1 | MIMAT0000181 |