Search
Search

| Assay Name: | ath-miR839-5p |
|---|---|
| miRBase Accession Number: | MI0005395 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACCAACCUUUCAUCGUUCCC |
| Species: | Arabidopsis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001143, MIR839 |
Mature miRNA Sequence: | UACCAACCUUUCAUCGUUCCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR839-5p | MIMAT0004261 |