Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12124
          Assay Name: hsa-miR-214-3p
          miRBase Accession Number: MI0000290
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 172138798 - 172138907 [-] on Build GRCh38
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Species: Human, Mouse, Anolis carolinensis, Artibeus jamaicensis, Bovine, Callithrix jacchus, Columba livia, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Ornithorhynchus anatinus, Otolemur garnettii, Papio hamadryas, Pteropus alecto, Python bivittatus, Rabbit, Taeniopygia guttata
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12124

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000062, mir-214

          Mature miRNA Sequence:

          ACAGCAGGCACAGACAGGCAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-214-3p MIMAT0000271
          Mouse mmu-miR-214-3p MIMAT0000661
          Anolis carolinensis aca-miR-214-3p MIMAT0021872
          Artibeus jamaicensis aja-miR-214 MIMAT0031144
          Bovine bta-miR-214 MIMAT0003825
          Callithrix jacchus cja-miR-214 MIMAT0039360
          View More Rows
          Columba livia cli-miR-214-3p MIMAT0038593
          Dasypus novemcinctus dno-miR-214-3p MIMAT0047759
          Daubentonia madagascariensis dma-miR-214 MIMAT0049272
          Dog cfa-miR-214 MIMAT0009847
          Guinea pig cpo-miR-214-3p MIMAT0047116
          Horse eca-miR-214 MIMAT0012963
          Microcebus murinus mmr-miR-214 MIMAT0049210
          Nomascus leucogenys nle-miR-214 MIMAT0049450
          Ornithorhynchus anatinus oan-miR-214-3p MIMAT0007171
          Otolemur garnettii oga-miR-214 MIMAT0049694
          Papio hamadryas pha-miR-214 MIMAT0049581
          Pteropus alecto pal-miR-214-3p MIMAT0040068
          Python bivittatus pbv-miR-214-3p MIMAT0039019
          Rabbit ocu-miR-214-3p MIMAT0048338
          Taeniopygia guttata tgu-miR-214-3p MIMAT0014603

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-214
          Stem-loop Accession # MI0000290
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location Chr. 1 - 172138798 - 172138907 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000271
          miRBase ID: hsa-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: Chr. 1 - 172138798 - 172138907 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-214
          Stem-loop Accession # MI0000698
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location Chr. 1 - 162223368 - 162223477 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000661
          miRBase ID: mmu-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: Chr. 1 - 162223368 - 162223477 [+] on Build GRCm38

          Anolis carolinensis

          Stem-loop ID aca-mir-214
          Stem-loop Accession # MI0018817
          Stem-loop Sequence
          CAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021872
          miRBase ID: aca-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Artibeus jamaicensis

          Stem-loop ID aja-mir-214
          Stem-loop Accession # MI0024366
          Stem-loop Sequence
          GGCCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031144
          miRBase ID: aja-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-214
          Stem-loop Accession # MI0005040
          Stem-loop Sequence
          GGCCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003825
          miRBase ID: bta-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-214
          Stem-loop Accession # MI0031874
          Stem-loop Sequence
          GUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039360
          miRBase ID: cja-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-214
          Stem-loop Accession # MI0030015
          Stem-loop Sequence
          GGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCGCAUGACAACCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038593
          miRBase ID: cli-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-214
          Stem-loop Accession # MI0039043
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047759
          miRBase ID: dno-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-214
          Stem-loop Accession # MI0040001
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049272
          miRBase ID: dma-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-214
          Stem-loop Accession # MI0010342
          Stem-loop Sequence
          GGCCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009847
          miRBase ID: cfa-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-214
          Stem-loop Accession # MI0038705
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047116
          miRBase ID: cpo-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-214
          Stem-loop Accession # MI0012714
          Stem-loop Sequence
          UCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012963
          miRBase ID: eca-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-214
          Stem-loop Accession # MI0039934
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049210
          miRBase ID: mmr-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-214
          Stem-loop Accession # MI0040199
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049450
          miRBase ID: nle-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-214
          Stem-loop Accession # MI0006889
          Stem-loop Sequence
          UGGGACUGGCUGGACGGAGUUGUCACGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCUGACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007171
          miRBase ID: oan-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-214
          Stem-loop Accession # MI0040478
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049694
          miRBase ID: oga-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-214
          Stem-loop Accession # MI0040356
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049581
          miRBase ID: pha-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-214
          Stem-loop Accession # MI0032476
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040068
          miRBase ID: pal-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-214
          Stem-loop Accession # MI0030287
          Stem-loop Sequence
          UGGAUAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAAUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039019
          miRBase ID: pbv-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-214
          Stem-loop Accession # MI0039348
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048338
          miRBase ID: ocu-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-214
          Stem-loop Accession # MI0013833
          Stem-loop Sequence
          GACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCGCAUGACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014603
          miRBase ID: tgu-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302971_pri
          TaqMan™ Pri-miRNA Assay : Mm03307122_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12124
          Ambion® Pre-miR™ miRNA Precursor : PM12124
          TaqMan™ MicroRNA Assay : 002306
          mirVana® miRNA mimic : MC12124
          TaqMan™ Advanced miRNA Assay : mmu481652_mir
          TaqMan™ Advanced miRNA Assay : 477974_mir


          miRBase Alias:

          Human: hsa-miR-214 (v17)
          Mouse: mmu-miR-214 (v17)
          Aeromonas caviae Ae398: aca-miR-214 (v18)
          Ochrobactrum anthropi: oan-miR-214 (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-214 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline