Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12302
          Assay Name: hsa-miR-216b-5p
          miRBase Accession Number: MI0005569
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 56000714 - 56000795 [-] on Build GRCh38
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Species: Human, Mouse, Rat, Anolis carolinensis, Bovine, Callithrix jacchus, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Guinea pig, Horse, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12302

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000054, mir-216

          Mature miRNA Sequence:

          AAAUCUCUGCAGGCAAAUGUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-216b-5p MIMAT0004959
          Mouse mmu-miR-216b-5p MIMAT0003729
          Rat rno-miR-216b-5p MIMAT0017846
          Anolis carolinensis aca-miR-216b-5p MIMAT0021876
          Bovine bta-miR-216b MIMAT0009266
          Callithrix jacchus cja-miR-216b MIMAT0039342
          View More Rows
          Chicken gga-miR-216b MIMAT0007748
          Columba livia cli-miR-216b-5p MIMAT0038598
          Cricetulus griseus cgr-miR-216b MIMAT0041157
          Dasypus novemcinctus dno-miR-216b-5p MIMAT0047764
          Dog cfa-miR-216b MIMAT0006592
          Guinea pig cpo-miR-216b-5p MIMAT0047121
          Horse eca-miR-216b MIMAT0013073
          Pan troglodytes ptr-miR-216b MIMAT0008072
          Pongo pygmaeus ppy-miR-216b MIMAT0015802
          Pteropus alecto pal-miR-216b-5p MIMAT0040051
          Python bivittatus pbv-miR-216b-5p MIMAT0039024
          Rhesus monkey mml-miR-216b MIMAT0006243

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-216b
          Stem-loop Accession # MI0005569
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAGGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACA
          Chromosome Location Chr. 2 - 56000714 - 56000795 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004959
          miRBase ID: hsa-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: Chr. 2 - 56000714 - 56000795 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-216b
          Stem-loop Accession # MI0004126
          Stem-loop Sequence
          UUGGCAGACUGGGAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAACCACACACUUACCUGUAGAGAUUCUUCAGUCUGACAA
          Chromosome Location Chr. 11 - 28746191 - 28746276 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003729
          miRBase ID: mmu-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: Chr. 11 - 28746191 - 28746276 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-216b
          Stem-loop Accession # MI0015435
          Stem-loop Sequence
          UGCACGUUGGCAGACUGGGAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAAUCACACACUUACUUGUAGAGAUUCUUCAGUCUGACAACAUCAAGCAU
          Chromosome Location Chr. 14 - 113101078 - 113101179 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017846
          miRBase ID: rno-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: Chr. 14 - 113101078 - 113101179 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-216b
          Stem-loop Accession # MI0018820
          Stem-loop Sequence
          UCAAAGUCACAGACUGAGAAAUCUCUGCAGGCAAAUGUGAUGUCUUGACCAGCAAUCUCACAGUUACCAGCAGAGGUUCUGCAAUCUGGACUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021876
          miRBase ID: aca-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-216b
          Stem-loop Accession # MI0009778
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009266
          miRBase ID: bta-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-216b
          Stem-loop Accession # MI0031855
          Stem-loop Sequence
          CUUCAAAGUGGCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAAUCACACAGUUACCCGCAGAGAUUCUACAGUCUGACACCUUCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039342
          miRBase ID: cja-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-216b
          Stem-loop Accession # MI0008205
          Stem-loop Sequence
          GAGUCACAGACUGGGAAAUCUCUGCAGGCAAAUGUGAUGUCUUUAUAGCAAUCUCACAAUUACCUGUAGAGAUUCUACAAUCUGGCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007748
          miRBase ID: gga-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-216b
          Stem-loop Accession # MI0030018
          Stem-loop Sequence
          GUCGCAGACUGGGAAAUCUCUGCAGGCAAAUGUGAUGUCUUUAUAGCAAUCUCACAAUUACCUGUAGAGAUUCUACAAUCUGGCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038598
          miRBase ID: cli-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-216b
          Stem-loop Accession # MI0033086
          Stem-loop Sequence
          GUGGCAGACUGAGAAAUCUCUGCAGGCAAAUGUGAUGUCAGUGAAGAAAUCACACACUUACCUGUAGAGAUUCUUCAGUCUGUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041157
          miRBase ID: cgr-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-216b
          Stem-loop Accession # MI0039046
          Stem-loop Sequence
          AAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCUAUAGAGAUUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047764
          miRBase ID: dno-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-216b
          Stem-loop Accession # MI0007987
          Stem-loop Sequence
          AAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCCGUAGAGAUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006592
          miRBase ID: cfa-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-216b
          Stem-loop Accession # MI0038708
          Stem-loop Sequence
          AAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAUCACACACUUACCUGUAAAGAUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047121
          miRBase ID: cpo-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-216b
          Stem-loop Accession # MI0012818
          Stem-loop Sequence
          GUGGCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013073
          miRBase ID: eca-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-216b
          Stem-loop Accession # MI0008582
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAGGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008072
          miRBase ID: ptr-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-216b
          Stem-loop Accession # MI0014867
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015802
          miRBase ID: ppy-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-216b
          Stem-loop Accession # MI0032468
          Stem-loop Sequence
          AAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCUGUAGAGAUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040051
          miRBase ID: pal-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-216b
          Stem-loop Accession # MI0030290
          Stem-loop Sequence
          GGUCCCAGUCACAGAUUGAGAAAUCUCUGCAGGCAAAUGUGAUGUCAUGAUUGCAAUCUCACAGUUACCAGUAGAGGUUCUGCAAUCUGGAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039024
          miRBase ID: pbv-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-216b
          Stem-loop Accession # MI0007673
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006243
          miRBase ID: mml-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307782_pri
          TaqMan™ Pri-miRNA Assay : Rn04229467_pri
          TaqMan™ Pri-miRNA Assay : Hs03305200_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12302
          Ambion® Pre-miR™ miRNA Precursor : PM12302
          TaqMan™ MicroRNA Assay : 002326
          mirVana® miRNA mimic : MC12302
          TaqMan™ Advanced miRNA Assay : mmu479456_mir
          TaqMan™ Advanced miRNA Assay : 479456_mir
          TaqMan™ Advanced miRNA Assay : rno479456_mir


          miRBase Alias:

          Human: hsa-miR-216b (v19)
          Mouse: mmu-miR-216b (v17)
          Aeromonas caviae Ae398: aca-miR-216b (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline