Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12682
          Assay Name: hsa-miR-202-5p
          miRBase Accession Number: MI0003130
          miRBase Version: v22.1
          Chromosome Location: Chr.10: 133247511 - 133247620 [-] on Build GRCh38
          Mature miRNA Sequence: UUCCUAUGCAUAUACUUCUUUG
          Species: Human, Chrysemys picta, Dog, Ophiophagus hannah, Python bivittatus
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12682

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000121, mir-202

          Mature miRNA Sequence:

          UUCCUAUGCAUAUACUUCUUUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-202-5p MIMAT0002810
          Chrysemys picta cpi-miR-202-5p MIMAT0037824
          Dog cfa-miR-202 MIMAT0009844
          Ophiophagus hannah oha-miR-202-5p MIMAT0036808
          Python bivittatus pbv-miR-202-5p MIMAT0039003

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-202
          Stem-loop Accession # MI0003130
          Stem-loop Sequence
          CGCCUCAGAGCCGCCCGCCGUUCCUUUUUCCUAUGCAUAUACUUCUUUGAGGAUCUGGCCUAAAGAGGUAUAGGGCAUGGGAAAACGGGGCGGUCGGGUCCUCCCCAGCG
          Chromosome Location Chr. 10 - 133247511 - 133247620 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002810
          miRBase ID: hsa-miR-202-5p
          Mature miRNA Sequence: UUCCUAUGCAUAUACUUCUUUG
          Chromosome Location: Chr. 10 - 133247511 - 133247620 [-] on Build GRCh38

          Chrysemys picta

          Stem-loop ID cpi-mir-202
          Stem-loop Accession # MI0029526
          Stem-loop Sequence
          GCUCGUUGUUCCUUUUUCCUAUGCAUAUACUUCUUUGAGAAUUGGAACUAAAGAGGCAUAGGGCAUGGGAAAAUGGGGCGACUGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037824
          miRBase ID: cpi-miR-202-5p
          Mature miRNA Sequence: UUCCUAUGCAUAUACUUCUUUG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-202
          Stem-loop Accession # MI0010339
          Stem-loop Sequence
          GUUCCUUUUUCCUAUGCAUAUACUUCUUUGAGGGUCUGGCCUGAAGAGGUGUAAGGCAUGGGAAAAGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009844
          miRBase ID: cfa-miR-202
          Mature miRNA Sequence: UUCCUAUGCAUAUACUUCUUUG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-202
          Stem-loop Accession # MI0031402
          Stem-loop Sequence
          UCGUCGUUCCUUUUUCCUAUGCAUAUACUUCUUUGAUAGGUCGAGUUUAAAGAGGCAUAAGGCAUGGGAAAAUGGGGCGAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036808
          miRBase ID: oha-miR-202-5p
          Mature miRNA Sequence: UUCCUAUGCAUAUACUUCUUUG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-202
          Stem-loop Accession # MI0030276
          Stem-loop Sequence
          UUCCUAUGCAUAUACUUCUUUGAGAAUUGAAUCUAAAGAGGCAUACGGCAUGGGAAAAUGGGGCGAUUGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039003
          miRBase ID: pbv-miR-202-5p
          Mature miRNA Sequence: UUCCUAUGCAUAUACUUCUUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303863_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12682
          Ambion® Pre-miR™ miRNA Precursor : PM12682
          TaqMan™ MicroRNA Assay : 002362
          mirVana® miRNA mimic : MC12682
          TaqMan™ Advanced miRNA Assay : 478755_mir


          miRBase Alias:

          Human: hsa-miR-202* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0