Search
Search

| Assay Name: | mmu-let-7g-3p |
|---|---|
| miRBase Accession Number: | MI0000137 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUGUACAGGCCACUGCCUUGC |
| Species: | Mouse, Ophiophagus hannah |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | ACUGUACAGGCCACUGCCUUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-let-7g-3p | MIMAT0004519 |
| Ophiophagus hannah | oha-let-7g-3p | MIMAT0036641 |