Search
Search

| Assay Name: | hsa-miR-523-3p |
|---|---|
| miRBase Accession Number: | MI0003153 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAACGCGCUUCCCUAUAGAGGGU |
| Species: | Human, Gorilla gorilla, Pan troglodytes |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | GAACGCGCUUCCCUAUAGAGGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-523-3p | MIMAT0002840 |
| Gorilla gorilla | ggo-miR-523-3p | MIMAT0036415 |
| Pan troglodytes | ptr-miR-523 | MIMAT0008207 |